siRNA Details
| Gene Symbol: 4930431P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045059.1 | 2 | 178 | |||
| GenBank mRNA | AK015279.1 | - | 2 | 179 | ||
| AK133263.1 | - | 2 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930431P03Rik| Gene Symbol: 4930431P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045059.1 | 2 | 178 | |||
| GenBank mRNA | AK015279.1 | - | 2 | 179 | ||
| AK133263.1 | - | 2 | 255 | |||