siRNA Details
| Gene Symbol: Lincenc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110430.1 | 3 | 1980 | |||
| NR_110431.1 | 2 | 1896 | ||||
| NR_110432.1 | 2 | 1942 | ||||
| GenBank mRNA | BC065997.1 | - | 2 | 1942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lincenc1| Gene Symbol: Lincenc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110430.1 | 3 | 1980 | |||
| NR_110431.1 | 2 | 1896 | ||||
| NR_110432.1 | 2 | 1942 | ||||
| GenBank mRNA | BC065997.1 | - | 2 | 1942 | ||