siRNA Details
| Gene Symbol: D630024D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102310.1 | 4 | 802 | |||
| NR_102311.1 | 4 | 757 | ||||
| GenBank mRNA | AK158188.1 | - | 4 | 803 | ||
| AK158233.1 | - | 4 | 740 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY595697.1 | - | 3 | 424 | Chr. 11 - 31818110 - 31823418 [-] on Build GRCm38 | |

D630024D03Rik
Genomic Map