siRNA Details
| Gene Symbol: 5133400J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110444.1 | 4 | 359 | |||
| GenBank mRNA | AK017220.1 | - | 4 | 359 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5133400J02Rik| Gene Symbol: 5133400J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110444.1 | 4 | 359 | |||
| GenBank mRNA | AK017220.1 | - | 4 | 359 | ||