siRNA Details
| Gene Symbol: LINC01186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110388.1 | 1 | 75 | |||
| GenBank mRNA | AL700385.1 | - | 1 | 43 | ||
| BC015977.1 | - | 1 | 48 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01186| Gene Symbol: LINC01186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110388.1 | 1 | 75 | |||
| GenBank mRNA | AL700385.1 | - | 1 | 43 | ||
| BC015977.1 | - | 1 | 48 | |||