siRNA Details
| Gene Symbol: LINC00944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033878.1 | 2 | 223 | |||
| GenBank mRNA | BC040060.1 | - | 2 | 189 | ||
| DA048939.1 | - | 4 | 516 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00944| Gene Symbol: LINC00944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033878.1 | 2 | 223 | |||
| GenBank mRNA | BC040060.1 | - | 2 | 189 | ||
| DA048939.1 | - | 4 | 516 | |||