siRNA Details
| Gene Symbol: C15orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302797.1 | NP_001289726.1 | 2 | 2447 | ||
| NM_207445.3 | NP_997328.1 | 2 | 2558 | |||
| GenBank mRNA | AK096850.1 | - | 2 | 2508 | ||
| BX647708.1 | - | 2 | 2545 | |||

C15orf54
Genomic Map