siRNA Details
| Gene Symbol: ZNF503-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024421.1 | 2 | 1027 | |||
| NR_024422.1 | 2 | 595 | ||||
| NR_110299.1 | 2 | 591 | ||||
| NR_110300.1 | 2 | 363 | ||||
| NR_110301.1 | 2 | 480 | ||||
| NR_110302.1 | 2 | 413 | ||||
| GenBank mRNA | BC017426.1 | - | 2 | 595 | ||
| BC029963.1 | - | 2 | 363 | |||
| DA634630.1 | - | 2 | 415 | |||

ZNF503-AS2
Genomic Map