This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC51145 SIRNA Assays ›
Gene Symbol:
LOC51145
Gene Name:
uncharacterized LOC51145
Gene Aliases:
-
Chromosome Location:
Chr.9: 125743754 - 125745929 on Build GRCh38
UniGene ID:
Hs.104671
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n545212

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC51145
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122032.1 1 244
GenBank mRNA AA397744.1 - 1 244
AF159054.1 - 1 867
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX095101.1 - 1 242 Chr. 9 - 125743770 - 125745927 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-nhgch:80/100.66.76.27:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline