siRNA Details
| Gene Symbol: LINC01445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120515.1 | 3 | 957 | |||
| NR_120516.1 | 3 | 490 | ||||
| GenBank mRNA | AV709400.1 | - | 3 | 419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01445| Gene Symbol: LINC01445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120515.1 | 3 | 957 | |||
| NR_120516.1 | 3 | 490 | ||||
| GenBank mRNA | AV709400.1 | - | 3 | 419 | ||