siRNA Details
| Gene Symbol: CDC37L1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121597.1 | 2 | 296 | |||
| NR_121598.1 | 2 | 218 | ||||
| GenBank mRNA | CK820219.1 | - | 2 | 209 | ||
| CR737367.1 | - | 2 | 295 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM263163.1 | - | 2 | 193 | Chr. 9 - 4676600 - 4679455 [-] on Build GRCh38 | |

CDC37L1-AS1
Genomic Map