This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LINC00310 SIRNA Assays ›
Gene Symbol:
LINC00310
Gene Name:
long intergenic non-protein coding RNA 310
Gene Aliases:
C21orf82, NCRNA00310
Chromosome Location:
Chr.21: 34180677 - 34189919 on Build GRCh38
UniGene ID:
Hs.720230
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n545752

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC00310
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027266.1 1 25
NR_027267.1 1 25
GenBank mRNA AF426258.1 - 1 25

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline