siRNA Details
| Gene Symbol: LINC00841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136148.1 | 2 | 396 | |||
| GenBank mRNA | AA432001.1 | - | 2 | 354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00841| Gene Symbol: LINC00841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136148.1 | 2 | 396 | |||
| GenBank mRNA | AA432001.1 | - | 2 | 354 | ||