siRNA Details
Gene Symbol: LINC01541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038325.1 | 6 | 1698 | |||
NR_038326.1 | 4 | 1313 | ||||
GenBank mRNA | AK093976.1 | - | 4 | 1298 | ||
BX648957.1 | - | 6 | 1699 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038325.1 | 6 | 1698 | |||
NR_038326.1 | 4 | 1313 | ||||
GenBank mRNA | AK093976.1 | - | 4 | 1298 | ||
BX648957.1 | - | 6 | 1699 |