siRNA Details
| Gene Symbol: LOC101929608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109878.1 | 5 | 913 | |||
| GenBank mRNA | AK310670.1 | - | 5 | 913 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929608| Gene Symbol: LOC101929608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109878.1 | 5 | 913 | |||
| GenBank mRNA | AK310670.1 | - | 5 | 913 | ||