siRNA Details
| Gene Symbol: LINC01018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024423.2 | 5 | 2799 | |||
| NR_024424.2 | 3 | 2813 | ||||
| GenBank mRNA | AK125283.1 | - | 5 | 2765 | ||
| DB572334.2 | - | 1 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01018| Gene Symbol: LINC01018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024423.2 | 5 | 2799 | |||
| NR_024424.2 | 3 | 2813 | ||||
| GenBank mRNA | AK125283.1 | - | 5 | 2765 | ||
| DB572334.2 | - | 1 | 473 | |||