siRNA Details
| Gene Symbol: ERVH48-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308491.1 | NP_001295420.1 | 2 | 735 | ||
| GenBank mRNA | AB610407.1 | - | 2 | 735 | ||
| BC005107.1 | - | 2 | 755 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERVH48-1| Gene Symbol: ERVH48-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308491.1 | NP_001295420.1 | 2 | 735 | ||
| GenBank mRNA | AB610407.1 | - | 2 | 735 | ||
| BC005107.1 | - | 2 | 755 | |||