siRNA Details
| Gene Symbol: LINC00927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033833.1 | 1 | 116 | |||
| GenBank mRNA | AK097782.1 | - | 87 | |||
| BC040586.1 | - | 1 | 79 | |||
| DB448280.1 | - | 1 | 116 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00927| Gene Symbol: LINC00927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033833.1 | 1 | 116 | |||
| GenBank mRNA | AK097782.1 | - | 87 | |||
| BC040586.1 | - | 1 | 79 | |||
| DB448280.1 | - | 1 | 116 | |||