siRNA Details
| Gene Symbol: SPATA41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028139.1 | 1 | 1570 | |||
| GenBank mRNA | AK057536.1 | - | 1 | 507 | ||
| AK124283.1 | - | 1 | 1565 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ710695.1 | - | 2 | 561 | Chr. 15 - 100348917 - 100350241 [-] on Build GRCh38 | |

SPATA41
Genomic Map