siRNA Details
| Gene Symbol: LOC285626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037889.1 | 1 | 498 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA733091.1 | - | 1 | 166 | Chr. 5 - 159331939 - 159416624 [+] on Build GRCh38 | |

LOC285626
Genomic Map