siRNA Details
| Gene Symbol: LOC101929084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110103.1 | 5 | 3033 | |||
| GenBank mRNA | BC041342.2 | - | 5 | 3033 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929084| Gene Symbol: LOC101929084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110103.1 | 5 | 3033 | |||
| GenBank mRNA | BC041342.2 | - | 5 | 3033 | ||