siRNA Details
| Gene Symbol: LOC101929125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109859.1 | 1 | 21 | |||
| GenBank mRNA | BC042140.1 | - | 1 | 21 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929125| Gene Symbol: LOC101929125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109859.1 | 1 | 21 | |||
| GenBank mRNA | BC042140.1 | - | 1 | 21 | ||