siRNA Details
| Gene Symbol: PRCAT47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131947.1 | 4 | 985 | |||
| NR_131948.1 | 3 | 871 | ||||
| GenBank mRNA | AK093002.1 | - | 3 | 871 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRCAT47| Gene Symbol: PRCAT47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131947.1 | 4 | 985 | |||
| NR_131948.1 | 3 | 871 | ||||
| GenBank mRNA | AK093002.1 | - | 3 | 871 | ||