siRNA Details
| Gene Symbol: LOC653712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034179.1 | 2 | 233 | |||
| GenBank mRNA | BX281830.1 | - | 2 | 233 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM558553.1 | - | 2 | 271 | Chr. 3 - 128861662 - 128871540 [-] on Build GRCh38 | |

LOC653712
Genomic Map