siRNA Details
| Gene Symbol: LINC00900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034148.1 | 3 | 684 | |||
| GenBank mRNA | AK090602.1 | - | 3 | 678 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499809.1 | - | 3 | 678 | Chr. 11 - 115754248 - 115760194 [-] on Build GRCh38 | |

LINC00900
Genomic Map