siRNA Details
| Gene Symbol: LINC00892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038461.1 | 3 | 675 | |||
| NR_038462.1 | 3 | 642 | ||||
| GenBank mRNA | BC042469.1 | - | 2 | 1706 | ||
| BX112139.1 | - | 3 | 445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00892| Gene Symbol: LINC00892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038461.1 | 3 | 675 | |||
| NR_038462.1 | 3 | 642 | ||||
| GenBank mRNA | BC042469.1 | - | 2 | 1706 | ||
| BX112139.1 | - | 3 | 445 | |||