This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2010002M12RIK SIRNA Assays ›
Gene Symbol:
2010002M12Rik
Gene Name:
RIKEN cDNA 2010002M12 gene
Gene Aliases:
AW412491, Ifit1b
Chromosome Location:
Chr.19: 34617051 - 34640743 on Build GRCm38
UniGene ID:
Mm.440727
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s100007

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2010002M12Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053217.2 NP_444447.1 3 1201
GenBank mRNA AK136525.1 - 2 1053
AK153247.1 - 3 1202
BC003493.1 - 1 555
BC138202.1 - 2 1032 8861221

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to virus
intracellular transport of viral protein in host cell
negative regulation of protein binding
positive regulation of viral genome replication
negative regulation of viral genome replication
negative regulation of defense response to virus by host
negative regulation of helicase activity
cellular response to type I interferon
cellular response to exogenous dsRNA
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rrztj:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline