This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DOK4 SIRNA Assays ›
Gene Symbol:
Dok4
Gene Name:
docking protein 4
Gene Aliases:
-
Chromosome Location:
Chr.8: 94863828 - 94876312 on Build GRCm38
UniGene ID:
Mm.203949
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s100101

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dok4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053246.3 NP_444476.1 2 367
GenBank mRNA AF418207.1 - 1 251
AK032352.1 - 2 357
AK043670.1 - 1 279
AK077612.1 - 2 358
AK142621.1 - 1 274
AK156485.1 - 2 367
BC004705.1 - 2 328 3499419

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108438, XM_008772316, XM_008772314, XM_008772315, XM_008772317, and others.

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
transmembrane receptor protein tyrosine kinase signaling pathway
nervous system development
intracellular signal transduction
receptor signaling protein activity
insulin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline