siRNA Details
| Gene Symbol: Spred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277256.1 | NP_001264185.1 | 3 | 937 | ||
| NM_033524.3 | NP_277059.1 | 3 | 937 | |||
| GenBank mRNA | AB063495.1 | - | 3 | 283 | ||
| AK155065.1 | - | 3 | 900 | |||
| AK155070.1 | - | 3 | 939 | |||
| BC041685.1 | - | 2 | 251 | |||
| BC057874.1 | - | 3 | 370 | 5319749 | ||
| BC079606.1 | - | 3 | 558 | 30355455 | ||

Spred1
Genomic Map