This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZFP287 SIRNA Assays ›
Gene Symbol:
Zfp287
Gene Name:
zinc finger protein 287
Gene Aliases:
B230333C16Rik, SKAT-2, Skat2, Znf287, mszf16, mszf74
Chromosome Location:
Chr.11: 62692444 - 62731900 on Build GRCm38
UniGene ID:
Mm.339982
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s100713

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp287
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133208.2 NP_573471.2 5 2492
GenBank mRNA AB010319.1 - 1 57
AF281141.1 - 5 2492
AK029572.1 - 5 2072
BC023790.1 - 3 1511
BC117876.1 - 5 2217 40095930
BC117877.1 - 5 2217 40095933

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of cytokine biosynthetic process
positive regulation of transcription, DNA-templated
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline