siRNA Details
| Gene Symbol: Bbc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133234.2 | NP_573497.1 | 4 | 805 | ||
| GenBank mRNA | AF332560.1 | - | 4 | 750 | ||
| BC044782.2 | - | 4 | 772 | |||
| BC060370.1 | - | 4 | 735 | |||
| BC147261.1 | - | 4 | 633 | 9056294 | ||
| BC147262.1 | - | 4 | 633 | 9056313 | ||

Bbc3
Genomic Map