siRNA Details
| Gene Symbol: Zfp286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138949.3 | NP_620399.3 | 6 | 1582 | ||
| GenBank mRNA | AK031547.1 | - | 6 | 1592 | ||
| AK173284.1 | - | 2 | 5241 | |||
| BC027023.1 | - | 2 | 1369 | 5365187 | ||
| BC050969.1 | - | 6 | 1772 | |||

Zfp286
Genomic Map