siRNA Details
| Gene Symbol: Zfp286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138949.3 | NP_620399.3 | 5 | 431 | ||
| GenBank mRNA | AK031547.1 | - | 5 | 441 | ||
| AK173284.1 | - | 1 | 4090 | |||
| BC027023.1 | - | 1 | 33 | 5365187 | ||
| BC038632.1 | - | 5 | 423 | |||
| BC050969.1 | - | 5 | 437 | |||

Zfp286
Genomic Map