siRNA Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148673.3 | NP_683514.2 | 17 | 1848 | ||
| NM_198936.1 | NP_945174.1 | 16 | 1769 | |||
| GenBank mRNA | AK029117.1 | - | 16 | 1739 | ||
| AK046262.1 | - | 16 | 1740 | |||
| AK049178.1 | - | 16 | 1738 | |||
| BC013810.1 | - | 14 | 1368 | |||
| BC023057.1 | - | 6 | 623 | |||

Slu7
Genomic Map