siRNA Details
| Gene Symbol: Tmprss11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033233.2 | NP_001028405.1 | 4 | 377 | ||
| GenBank mRNA | AK142820.1 | - | 4 | 378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11a| Gene Symbol: Tmprss11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033233.2 | NP_001028405.1 | 4 | 377 | ||
| GenBank mRNA | AK142820.1 | - | 4 | 378 | ||