This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BZRAP1 SIRNA Assays ›
Gene Symbol:
Bzrap1
Gene Name:
benzodiazepine receptor associated protein 1
Gene Aliases:
D230016K05, PRAX-1, mKIAA0612
Chromosome Location:
Chr.11: 87759066 - 87785928 on Build GRCm38
UniGene ID:
Mm.342019
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s101703

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bzrap1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172449.2 NP_766037.2 29 6250
GenBank mRNA AK051895.1 - 8 2199
AK122329.1 - 7 1121
BC055691.1 - 9 1001
BC066146.1 - 8 1261 30470084
BC083181.1 - 10 1101

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
benzodiazepine receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-b4tm8:80/100.66.75.155:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline