This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other THSD7B SIRNA Assays ›
Gene Symbol:
Thsd7b
Gene Name:
thrombospondin, type I, domain containing 7B
Gene Aliases:
1700074E13Rik, B930082A18, D130067I03Rik
Chromosome Location:
Chr.1: 129273304 - 130219278 on Build GRCm38
UniGene ID:
Mm.380518
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s102252

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Thsd7b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172485.3 NP_766073.2 21 4154
GenBank mRNA AK166575.1 - 9 1338
BC063250.1 - 21 3984 6410178

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001080427, AB051466.
This siRNA also targets the following transcripts in Rat: NM_001191669, XM_008769435.

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline