siRNA Details
| Gene Symbol: Lemd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033250.4 | NP_001028422.2 | 3 | 240 | ||
| GenBank mRNA | AK016010.1 | - | 3 | 245 | ||
| AK144269.1 | - | 247 | ||||
| BC048759.1 | - | 3 | 204 | |||
| BC115462.1 | - | 3 | 221 | 40100287 | ||
| BC115463.1 | - | 3 | 221 | 40100292 | ||

Lemd1
Genomic Map