This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZFP598 SIRNA Assays ›
Gene Symbol:
Zfp598
Gene Name:
zinc finger protein 598
Gene Aliases:
BC023040, Ntrap, Znf598
Chromosome Location:
Chr.17: 24669693 - 24682016 on Build GRCm38
UniGene ID:
Mm.219581
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s102939

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp598
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183149.1 NP_898972.1 11 2321
GenBank mRNA AK131127.1 - 12 2256
AK138860.1 - 8 2434
BC023040.1 - 4 223
BC024690.1 - 11 2106
BC038138.1 - 1 83
BC050859.1 - 11 2321 6328878

Back To Top

More Information


Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
zinc ion binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xg9l8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline