This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM4788 SIRNA Assays ›
Gene Symbol:
Gm4788
Gene Name:
predicted gene 4788
Gene Aliases:
Cfhrc, EG214403
Chromosome Location:
Chr.1: 139697919 - 139781239 on Build GRCm38
UniGene ID:
Mm.337613
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s103077

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm4788
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029977.3 NP_001025148.3 15 2555
NM_001160303.1 NP_001153775.1 15 2552
NM_001160304.1 NP_001153776.1 14 2378
GenBank mRNA AJ871401.1 - 15 2484
AJ871402.1 - 14 2307
BC141136.1 - 15 2524 9055748
BC145444.1 - 15 2524 9054002
M29009.1 - 14 2339

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0