This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC39A2 SIRNA Assays ›
Gene Symbol:
Slc39a2
Gene Name:
solute carrier family 39 (zinc transporter), member 2
Gene Aliases:
F730005G13Rik, Gm1789, Gm289, Zip2
Chromosome Location:
Chr.14: 51891972 - 51896745 on Build GRCm38
UniGene ID:
Mm.281343
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s103209

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc39a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039676.2 NP_001034765.2 2 370
GenBank mRNA AK089322.1 - 2 322
AK170178.1 - 2 325
BC113769.1 - 2 340 40057862
BY200549.1 - 2 372

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107260, XM_008770521.

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

zinc II ion transport
zinc II ion transmembrane transport
zinc ion transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline