siRNA Details
| Gene Symbol: Rap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024457.2 | NP_077777.1 | 6 | 590 | ||
| GenBank mRNA | AK143910.1 | - | 6 | 573 | ||
| AK153870.1 | - | 6 | 547 | |||
| AK163242.1 | - | 6 | 654 | |||
| AK167548.1 | - | 6 | 546 | |||
| AK205627.1 | - | 2 | 119 | |||
| BC033382.1 | - | 6 | 530 | |||
| BC052480.1 | - | 6 | 590 | |||
| BC094066.1 | - | 6 | 530 | |||
| BC147390.1 | - | 6 | 462 | 9056464 | ||
| BC147391.1 | - | 6 | 462 | 9056441 | ||
| BC147700.1 | - | 5 | 448 | 9087459 | ||
| BC147713.1 | - | 5 | 448 | 9087472 | ||
| CK793274.1 | - | 1 | 107 | |||

Rap1b
Genomic Map