This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other WDR18 SIRNA Assays ›
Gene Symbol:
Wdr18
Gene Name:
WD repeat domain 18
Gene Aliases:
2310012I10Rik, AU044733, AW122032
Chromosome Location:
Chr.10: 79960152 - 79969246 on Build GRCm38
UniGene ID:
Mm.35828
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s103518

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wdr18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175450.4 NP_780659.2 2 273
GenBank mRNA AK032251.1 - 2 271
AK050106.1 - 2 271
AK082695.1 - 2 275
AK087505.1 - 1 904
AK133716.1 - 2 274
AK160713.1 - 2 271
AK163465.1 - 1 901
AK166113.1 - 271
AK168691.1 - 2 271
BC095984.1 - 2 273 4978914

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline