siRNA Details
| Gene Symbol: Nol10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008421.1 | NP_001008421.1 | 20 | 1952 | ||
| GenBank mRNA | AK148210.1 | - | 20 | 1968 | ||
| AK160864.1 | - | 20 | 1988 | |||
| AK166160.1 | - | 20 | 1982 | |||
| BC086676.1 | - | 20 | 1952 | 30659737 | ||

Nol10
Genomic Map