This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MARF1 SIRNA Assays ›
Gene Symbol:
Marf1
Gene Name:
meiosis arrest female 1
Gene Aliases:
4921513D23Rik, BC031575, C87306, E030019O05, Lkap, mKIAA0430
Chromosome Location:
Chr.16: 14109166 - 14159274 on Build GRCm38
UniGene ID:
Mm.394072
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s104692

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Marf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081154.2 NP_001074623.1 8 1843
GenBank mRNA AK035704.1 - 1497
AK220341.1 - 5 3507
BC031575.1 - 1 15
BC069852.1 - 4 529

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
female meiotic division
regulation of gene expression
negative regulation of phosphatase activity
cell differentiation
oogenesis
meiotic cell cycle
nucleotide binding
molecular_function
nucleic acid binding
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline