This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VMN2R108 SIRNA Assays ›
Gene Symbol:
Vmn2r108 Vmn2r110
Gene Name:
vomeronasal 2, receptor 108
vomeronasal 2, receptor 110
Gene Aliases:
EG627805
EG224582
Chromosome Location:
Chr.17: 20462373 - 20481236 on Build GRCm38
Chromosome Location:
Chr.17: 20573829 - 20596259 on Build GRCm38
UniGene ID:
Mm.469853
Mm.469851
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s104836

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vmn2r108
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104570.1 NP_001098040.1 3 1055
Gene Symbol: Vmn2r110
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104572.1 NP_001098042.1 3 1055

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008759288.

Set Membership:

G-protein coupled receptor

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0