This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other D17WSU92E SIRNA Assays ›
Gene Symbol:
D17Wsu92e
Gene Name:
DNA segment, Chr 17, Wayne State University 92, expressed
Gene Aliases:
AU020189, C030010A15, C80239
Chromosome Location:
Chr.17: 27751232 - 27820542 on Build GRCm38
UniGene ID:
Mm.196577
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s104862

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: D17Wsu92e
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033279.3 NP_001028451.2 4 784
NM_001044719.2 NP_001038184.1 4 784
GenBank mRNA AK143016.1 - 4 696
AK161603.1 - 4 785
AK165011.1 - 4 784
BC132324.1 - 5 710 40130601
BC132326.1 - 5 710 40130603
BC137573.1 - 4 784 8860592
BC137576.1 - 4 784 8860595
BC145403.1 - 4 784 9053961
BU922193.1 - 3 382
CF536829.1 - 1 37

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_022758, NM_024294, BC010184, BC075810, AK056964, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
macroautophagy
molecular_function
ubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline