siRNA Details
| Gene Symbol: Cox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144874.4 | NP_659123.2 | 5 | 709 | ||
| GenBank mRNA | AK048099.1 | - | 5 | 691 | ||
| AK051792.1 | - | 5 | 607 | |||
| AK141631.1 | - | 5 | 641 | |||
| BC011509.1 | - | 5 | 669 | 3981950 | ||
| BC021498.1 | - | 5 | 629 | |||

Cox15
Genomic Map