siRNA Details
| Gene Symbol: Proser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159657.1 | NP_001153129.1 | 2, 3 | 509 | ||
| NM_144883.4 | NP_659132.2 | 2, 3 | 363 | |||
| GenBank mRNA | AI317549.1 | - | 2, 3 | 371 | ||
| AK077380.1 | - | 2, 3 | 339 | |||
| AK132370.1 | - | 313 | ||||
| BB569558.2 | - | 2, 3 | 338 | |||
| BC023193.1 | - | 1, 2 | 222 | 4946741 | ||
| BC025867.1 | - | 3, 4 | 470 | 5052194 | ||
| BC029968.1 | - | 2, 3 | 326 | |||

Proser2
Genomic Map