This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LRSAM1 SIRNA Assays ›
Gene Symbol:
Lrsam1
Gene Name:
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 32925215 - 32961654 on Build GRCm38
UniGene ID:
Mm.248037
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s105793

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrsam1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199302.2 NP_955006.1 14, 15 1333
GenBank mRNA BC049146.1 - 14, 15 1329 6306042

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
transport
protein transport
protein catabolic process
negative regulation of endocytosis
viral budding
protein autoubiquitination
ubiquitin-dependent endocytosis
ubiquitin-protein transferase activity
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-mbm4f:80/100.66.76.81:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline